Anti-IL17RB Mouse Monoclonal Antibody [clone: 170220]

Supplier: R & D Systems
MAB1207
220018-896EA 763.57 USD
220018-896
Anti-IL17RB Mouse Monoclonal Antibody [clone: 170220]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 170220
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR