Anti-FGF3 Mouse Monoclonal Antibody [clone: 254625]

Supplier: R & D Systems
MAB1206
220018-895EA 763.57 USD
220018-895
Anti-FGF3 Mouse Monoclonal Antibody [clone: 254625]
Antibodies
Type: Primary
Antigen Symbol: FGF3
Antigen Name: fibroblast growth factor 3
Antigen Synonyms: HBGF-3, INT2
Clonality: Monoclonal
Clone: 254625
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR