Anti-JNK1 Rabbit Monoclonal Antibody [clone: 1006A]

Supplier: R & D Systems
MAB1205
102885-248EA 633.42 USD
102885-248
Anti-JNK1 Rabbit Monoclonal Antibody [clone: 1006A]
Antibodies
Simple Western

Type: Primary
Antigen: JNK1
Clonality: Monoclonal
Clone: 1006A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR