Anti-IL20 Rat Monoclonal Antibody [clone: 380605]

Supplier: R & D Systems
MAB12043
220018-894EA 763.57 USD
220018-894
Anti-IL20 Rat Monoclonal Antibody [clone: 380605]
Antibodies
Type: Primary
Antigen: IL20
Clonality: Monoclonal
Clone: 380605
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR