Anti-IL20 Rat Monoclonal Antibody [clone: 176005]

Supplier: R & D Systems
MAB1204
220018-892EA 763.57 USD
220018-892
Anti-IL20 Rat Monoclonal Antibody [clone: 176005]
Antibodies
Type: Primary
Antigen: IL20
Clonality: Monoclonal
Clone: 176005
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR