Anti-INHBA Mouse Monoclonal Antibody [clone: 132815]

Supplier: R & D Systems
MAB1203
220018-891EA 772.25 USD
220018-891
Anti-INHBA Mouse Monoclonal Antibody [clone: 132815]
Antibodies
Type: Primary
Antigen: INHBA
Clonality: Monoclonal
Clone: 132815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR