Anti-CST5 Mouse Monoclonal Antibody [clone: 206006]

Supplier: R & D Systems
MAB1202
220018-890EA 763.57 USD
220018-890
Anti-CST5 Mouse Monoclonal Antibody [clone: 206006]
Antibodies
Type: Primary
Antigen: CST5
Clonality: Monoclonal
Clone: 206006
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR