Anti-CST2 Mouse Monoclonal Antibody [clone: 202332]

Supplier: R & D Systems
MAB1201
220018-889EA 763.57 USD
220018-889
Anti-CST2 Mouse Monoclonal Antibody [clone: 202332]
Antibodies
Type: Primary
Antigen: CST2
Clonality: Monoclonal
Clone: 202332
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR