Anti-TNFRSF12A Rat Monoclonal Antibody [clone: 314502]

Supplier: R & D Systems
MAB1199
220018-888EA 594.37 USD
220018-888
Anti-TNFRSF12A Rat Monoclonal Antibody [clone: 314502]
Antibodies
Type: Primary
Antigen: TNFRSF12A
Clonality: Monoclonal
Clone: 314502
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR