Anti-LGALS3 Rat Monoclonal Antibody [clone: 202213]

Supplier: R & D Systems
MAB1197
220018-887EA 779.48 USD
220018-887
Anti-LGALS3 Rat Monoclonal Antibody [clone: 202213]
Antibodies
Simple Western

Type: Primary
Antigen: LGALS3
Clonality: Monoclonal
Clone: 202213
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR