Anti-IL18BP Mouse Monoclonal Antibody [clone: 136031]

Supplier: R & D Systems
MAB1192
220018-883EA 779.48 USD
220018-883
Anti-IL18BP Mouse Monoclonal Antibody [clone: 136031]
Antibodies
Type: Primary
Antigen: IL18BP
Clonality: Monoclonal
Clone: 136031
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR