Anti-NOTCH2 Mouse Monoclonal Antibody [clone: 199509]

Supplier: R & D Systems
MAB1190
220018-881EA 763.57 USD
220018-881
Anti-NOTCH2 Mouse Monoclonal Antibody [clone: 199509]
Antibodies
Type: Primary
Antigen: NOTCH2
Clonality: Monoclonal
Clone: 199509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR