Anti-IL18BP Mouse Monoclonal Antibody [clone: 136033]

Supplier: R & D Systems
MAB119
220018-880EA 772.25 USD
220018-880
Anti-IL18BP Mouse Monoclonal Antibody [clone: 136033]
Antibodies
Type: Primary
Antigen: IL18BP
Clonality: Monoclonal
Clone: 136033
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR