Anti-JAM3 Mouse Monoclonal Antibody [clone: 208206]

Supplier: R & D Systems
MAB1189
220018-879EA 772.25 USD
220018-879
Anti-JAM3 Mouse Monoclonal Antibody [clone: 208206]
Antibodies
Type: Primary
Antigen: JAM3
Clonality: Monoclonal
Clone: 208206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR