Anti-CD300A Rat Monoclonal Antibody [clone: 172224]

Supplier: R & D Systems
MAB1186
220018-877EA 594.37 USD
220018-877
Anti-CD300A Rat Monoclonal Antibody [clone: 172224]
Antibodies
Type: Primary
Antigen: CD300A
Clonality: Monoclonal
Clone: 172224
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR