Anti-AHSG Mouse Monoclonal Antibody [clone: 162922]

Supplier: R & D Systems
MAB1184
220018-875EA 772.25 USD
220018-875
Anti-AHSG Mouse Monoclonal Antibody [clone: 162922]
Antibodies
Type: Primary
Antigen: AHSG
Clonality: Monoclonal
Clone: 162922
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR