Anti-CTSS Rat Monoclonal Antibody [clone: 248718]

Supplier: R & D Systems
MAB1183
220018-874EA 772.25 USD
220018-874
Anti-CTSS Rat Monoclonal Antibody [clone: 248718]
Antibodies
Type: Primary
Antigen: CTSS
Clonality: Monoclonal
Clone: 248718
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR