Anti-MME Mouse Monoclonal Antibody [clone: 715820]

Supplier: R & D Systems
MAB11821
102885-246EA 577.02 USD
102885-246
Anti-MME Mouse Monoclonal Antibody [clone: 715820]
Antibodies
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: 715820
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR