Anti-MME Mouse Monoclonal Antibody [clone: 212504]

Supplier: R & D Systems
MAB1182
220018-873EA 763.57 USD
220018-873
Anti-MME Mouse Monoclonal Antibody [clone: 212504]
Antibodies
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: 212504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR