Anti-AGER Rat Monoclonal Antibody [clone: 69723]

Supplier: R & D Systems
MAB11795
102885-244EA 501.82 USD
102885-244
Anti-AGER Rat Monoclonal Antibody [clone: 69723]
Antibodies
Type: Primary
Antigen: AGER
Clonality: Monoclonal
Clone: 69723
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR