Anti-AGER Rat Monoclonal Antibody [clone: 6975]

Supplier: R & D Systems
MAB11794
102885-242EA 594.37 USD
102885-242
Anti-AGER Rat Monoclonal Antibody [clone: 6975]
Antibodies
Type: Primary
Antigen: AGER
Clonality: Monoclonal
Clone: 6975
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR