Anti-AGER Rat Monoclonal Antibody [clone: 17549]

Supplier: R & D Systems
MAB11792
220018-869EA 772.25 USD
220018-869
Anti-AGER Rat Monoclonal Antibody [clone: 17549]
Antibodies
Type: Primary
Antigen: AGER
Clonality: Monoclonal
Clone: 17549
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR