Anti-IL20RA Mouse Monoclonal Antibody [clone: 173707]

Supplier: R & D Systems
MAB11761
220018-865EA 772.25 USD
220018-865
Anti-IL20RA Mouse Monoclonal Antibody [clone: 173707]
Antibodies
Type: Primary
Antigen: IL20RA
Clonality: Monoclonal
Clone: 173707
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR