Anti-IL20RA Mouse Monoclonal Antibody [clone: 173722]

Supplier: R & D Systems
MAB1176
220018-864EA 763.57 USD
220018-864
Anti-IL20RA Mouse Monoclonal Antibody [clone: 173722]
Antibodies
Type: Primary
Antigen: IL20RA
Clonality: Monoclonal
Clone: 173722
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR