Anti-CTSO Mouse Monoclonal Antibody [clone: 180406]

Supplier: R & D Systems
MAB1174
220018-863EA 763.57 USD
220018-863
Anti-CTSO Mouse Monoclonal Antibody [clone: 180406]
Antibodies
Type: Primary
Antigen: CTSO
Clonality: Monoclonal
Clone: 180406
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR