Anti-ICAM5 Rat Monoclonal Antibody [clone: 191321]

Supplier: R & D Systems
MAB1173
220018-862EA 763.57 USD
220018-862
Anti-ICAM5 Rat Monoclonal Antibody [clone: 191321]
Antibodies
Type: Primary
Antigen: ICAM5
Clonality: Monoclonal
Clone: 191321
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR