Anti-IL-12/IL-35 p35 Mouse Monoclonal Antibody [clone: 116219]

Supplier: R & D Systems
MAB1171
220018-860EA 772.25 USD
220018-860
Anti-IL-12/IL-35 p35 Mouse Monoclonal Antibody [clone: 116219]
Antibodies
Type: Primary
Antigen: IL-12/IL-35 p35
Clonality: Monoclonal
Clone: 116219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR