Anti-SFRP2 Rat Monoclonal Antibody [clone: 331022]

Supplier: R & D Systems
MAB1169
220018-859EA 594.37 USD
220018-859
Anti-SFRP2 Rat Monoclonal Antibody [clone: 331022]
Antibodies
Type: Primary
Antigen: SFRP2
Clonality: Monoclonal
Clone: 331022
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR