Anti-PRLR Mouse Monoclonal Antibody [clone: 250448]

Supplier: R & D Systems
MAB1167
220018-858EA 772.25 USD
220018-858
Anti-PRLR Mouse Monoclonal Antibody [clone: 250448]
Antibodies
Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: 250448
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR