Anti-LGALS3 Mouse Monoclonal Antibody [clone: 194801]

Supplier: R & D Systems
MAB11542
102885-238EA 616.06 USD
102885-238
Anti-LGALS3 Mouse Monoclonal Antibody [clone: 194801]
Antibodies
Type: Primary
Antigen: LGALS3
Clonality: Monoclonal
Clone: 194801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR