Anti-LGALS3 Mouse Monoclonal Antibody [clone: 194804]

Supplier: R & D Systems
MAB1154
220018-852EA 763.57 USD
220018-852
Anti-LGALS3 Mouse Monoclonal Antibody [clone: 194804]
Antibodies
Type: Primary
Antigen: LGALS3
Clonality: Monoclonal
Clone: 194804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR