Anti-MMP25 Mouse Monoclonal Antibody [clone: 141811]

Supplier: R & D Systems
MAB11422
220018-845EA 763.57 USD
220018-845
Anti-MMP25 Mouse Monoclonal Antibody [clone: 141811]
Antibodies
Type: Primary
Antigen: MMP25
Clonality: Monoclonal
Clone: 141811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR