Anti-MMP25 Mouse Monoclonal Antibody [clone: 141808]

Supplier: R & D Systems
MAB1142
220018-843EA 763.57 USD
220018-843
Anti-MMP25 Mouse Monoclonal Antibody [clone: 141808]
Antibodies
Type: Primary
Antigen: MMP25
Clonality: Monoclonal
Clone: 141808
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR