Anti-SPINT1 Rat Monoclonal Antibody [clone: 199732]

Supplier: R & D Systems
MAB1141
220018-842EA 763.57 USD
220018-842
Anti-SPINT1 Rat Monoclonal Antibody [clone: 199732]
Antibodies
Type: Primary
Antigen: SPINT1
Clonality: Monoclonal
Clone: 199732
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR