Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: 194211]

Supplier: R & D Systems
MAB11381
102885-236EA 594.37 USD
102885-236
Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: 194211]
Antibodies
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 194211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR