Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: 194212]

Supplier: R & D Systems
MAB1138
220018-839EA 779.48 USD
220018-839
Anti-SIGLEC7 Mouse Monoclonal Antibody [clone: 194212]
Antibodies
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 194212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR