Anti-NTN4 Rat Monoclonal Antibody [clone: 299612]

Supplier: R & D Systems
MAB1132
220018-836EA 566.89 USD
220018-836
Anti-NTN4 Rat Monoclonal Antibody [clone: 299612]
Antibodies
Type: Primary
Antigen: NTN4
Clonality: Monoclonal
Clone: 299612
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR