Anti-ERBB4 Mouse Monoclonal Antibody [clone: 182803]

Supplier: R & D Systems
MAB1131
220018-834EA 763.57 USD
220018-834
Anti-ERBB4 Mouse Monoclonal Antibody [clone: 182803]
Antibodies
Type: Primary
Antigen: ERBB4
Clonality: Monoclonal
Clone: 182803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR