Anti-CTSE Rat Monoclonal Antibody [clone: 186513]

Supplier: R & D Systems
MAB1130
220018-833EA 763.57 USD
220018-833
Anti-CTSE Rat Monoclonal Antibody [clone: 186513]
Antibodies
Type: Primary
Antigen: CTSE
Clonality: Monoclonal
Clone: 186513
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR