Anti-BMP-2/-4 Mouse Monoclonal Antibody [clone: 169125]

Supplier: R & D Systems
MAB1128
220018-830EA 763.57 USD
220018-830
Anti-BMP-2/-4 Mouse Monoclonal Antibody [clone: 169125]
Antibodies
Type: Primary
Antigen Symbol: BMP-2/-4
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 169125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR