Anti-EPGN Rat Monoclonal Antibody [clone: 187204]

Supplier: R & D Systems
MAB11271
220018-829EA 763.57 USD
220018-829
Anti-EPGN Rat Monoclonal Antibody [clone: 187204]
Antibodies
Type: Primary
Antigen: EPGN
Clonality: Monoclonal
Clone: 187204
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR