Anti-MME Rat Monoclonal Antibody [clone: 200103]

Supplier: R & D Systems
MAB1126
220018-827EA 763.57 USD
220018-827
Anti-MME Rat Monoclonal Antibody [clone: 200103]
Antibodies
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: 200103
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR