Anti-LY76 Rat Monoclonal Antibody [clone: TER-119]

Supplier: R & D Systems
MAB1125
220018-825EA 763.57 USD
220018-825
Anti-LY76 Rat Monoclonal Antibody [clone: TER-119]
Antibodies
Type: Primary
Antigen: LY76
Clonality: Monoclonal
Clone: TER-119
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR