Anti-CTSA Rat Monoclonal Antibody [clone: 191424]

Supplier: R & D Systems
MAB11231
220018-823EA 763.57 USD
220018-823
Anti-CTSA Rat Monoclonal Antibody [clone: 191424]
Antibodies
Type: Primary
Antigen: CTSA
Clonality: Monoclonal
Clone: 191424
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR