Anti-TACSTD2 Rat Monoclonal Antibody [clone: 168215]

Supplier: R & D Systems
MAB1122
220018-821EA 772.25 USD
220018-821
Anti-TACSTD2 Rat Monoclonal Antibody [clone: 168215]
Antibodies
Type: Primary
Antigen: TACSTD2
Clonality: Monoclonal
Clone: 168215
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR