Anti-TNFSF10 Rat Monoclonal Antibody [clone: 170533]

Supplier: R & D Systems
MAB1121
220018-820EA 772.25 USD
220018-820
Anti-TNFSF10 Rat Monoclonal Antibody [clone: 170533]
Antibodies
Type: Primary
Antigen: TNFSF10
Clonality: Monoclonal
Clone: 170533
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR