Anti-FZD1 Rat Monoclonal Antibody [clone: 162531]

Supplier: R & D Systems
MAB11201-050 MAB11201-100
220018-816EA 339.85 USD
220018-816 220018-817
Anti-FZD1 Rat Monoclonal Antibody [clone: 162531]
Antibodies
Type: Primary
Antigen Symbol: FZD1
Antigen Name: frizzled class receptor 1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 162531
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR