Anti-FZD1 Rat Monoclonal Antibody [clone: 162526]

Supplier: R & D Systems
MAB1120-500 MAB1120-050
220018-815EA 339.85 USD
220018-815 220018-819
Anti-FZD1 Rat Monoclonal Antibody [clone: 162526]
Antibodies
Type: Primary
Antigen Symbol: FZD1
Antigen Name: frizzled homolog 1 (Drosophila)
Antigen Synonyms: AW227548, FZ-1, Fz1
Clonality: Monoclonal
Clone: 162526
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR