Anti-DKK3 Mouse Monoclonal Antibody [clone: 138728]

Supplier: R & D Systems
MAB11181
220018-811EA 779.48 USD
220018-811
Anti-DKK3 Mouse Monoclonal Antibody [clone: 138728]
Antibodies
Type: Primary
Antigen: DKK3
Clonality: Monoclonal
Clone: 138728
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR