Anti-DKK3 Mouse Monoclonal Antibody [clone: 138726]

Supplier: R & D Systems
MAB1118-050 MAB1118-500
220018-810EA 339.85 USD
220018-810 220018-812
Anti-DKK3 Mouse Monoclonal Antibody [clone: 138726]
Antibodies
Type: Primary
Antigen: DKK3
Clonality: Monoclonal
Clone: 138726
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR